missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-Ibrutinib-d4, TRC

Product Code. 30335841
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335841 25 mg 25mg
30331592 5 mg 5mg
30340687 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335841 Supplier Toronto Research Chemicals Supplier No. TRCB65671725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-Ibrutinib-d4
Molecular Formula C27 D4 H26 N6 O3
Quantity 25 mg
Synonym (3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester-d4, (R)-Butyl 3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidine-1-carboxylate-d4
InChI Formula InChI=1S/C27H30N6O3/c1-27(2,3)36-26(34)32-15-7-8-19(16-32)33-25-22(24(28)29-17-30-25)23(31-33)18-11-13-21(14-12-18)35-20-9-5-4-6-10-20/h4-6,9-14,17,19H,7-8,15-16H2,1-3H3,(H2,28,29,30)/t19-/m1/s1/i15D2,16D2
SMILES [2H]C1([2H])CC[C@@H](n2nc(c3ccc(Oc4ccccc4)cc3)c5c(N)ncnc25)C([2H])([2H])N1C(=O)OC(C)(C)C
IUPAC Name tert-butyl (3R)-3-[4-amino-3-(4-phenoxyphenyl)pyrazolo[3,4-d]pyrimidin-1-yl]-2,2,6,6-tetradeuteriopiperidine-1-carboxylate
Molecular Weight (g/mol) 490.59
Formula Weight 490.263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.