Learn More
N-Boc-5-oxo-L-proline, 97%
Product Code. 15495068
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495068 5 g 5g
15405078 250 mg 250mg
15485068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15495068 Supplier Thermo Scientific Alfa Aesar Supplier No. H58884.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 53100-44-0 | C10H15NO5 | 229.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53100-44-0
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD00672316
InChI Key MJLQPFJGZTYCMH-LURJTMIESA-N
Synonym boc-pyr-oh, n-boc-l-pyroglutamic acid, s-boc-5-oxopyrrolidine-2-carboxylic acid, boc-l-pyroglutamic acid, 2s-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylic acid, 5-oxo-, 1-1,1-dimethylethyl ester, 2s, 1-tert-butoxycarbonyl-5-oxo-l-proline, bocpyr, 2s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid
PubChem CID 104412
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(O)=O
Specifications
Melting Point 118°C to 123°C
Quantity 5 g
Sensitivity Moisture sensitive
Optical Rotation −35° (c=1 in Chloroform)
Formula Weight 229.23
Percent Purity 97%
Chemical Name or Material N-Boc-5-oxo-L-proline