missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-chloro-L-phenylalanine, 95%, Thermo Scientific™

Product Code. 15467798
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15467798 25 g 25g
15447798 1 g 1g
15457798 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15467798

Brand: Thermo Scientific Alfa Aesar H61283.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68090-88-0
Molecular Formula C14H17ClNO4
Molecular Weight (g/mol) 298.74
MDL Number MFCD00057808
InChI Key BETBOAZCLSJOBQ-NSHDSACASA-M
Synonym boc-phe 4-cl-oh, boc-l-4-chlorophenylalanine, boc-4-chloro-l-phenylalanine, boc-l-4-chlorophe, boc-l-phe 4-cl-oh, n-boc-4-chloro-l-phenylalanine, n-tert-butoxycarbonyl-4-chloro-l-phenylalanine, s-n-boc-4-chlorophenylalanine, s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, boc-p-chloro-phe-oh
PubChem CID 2734499
IUPAC Name (2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C=C1)C([O-])=O

Specifications

Melting Point ∼110°C
Quantity 25 g
Beilstein 4261946
Formula Weight 299.75
Percent Purity 95%
Chemical Name or Material N-Boc-4-chloro-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.