Learn More
N-Boc-4-bromo-L-phenylalanine, 98%
Product Code. 15498427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498427 1 g 1g
15408437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15498427 Supplier Thermo Scientific Alfa Aesar Supplier No. H51969.03
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
CAS: 62129-39-9 | C14H17BrNO4 | 343.20 g/mol
N-Boc-4-bromo-L-phenylalanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-4-bromo-L-phenylalanine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62129-39-9
Molecular Formula C14H17BrNO4
Molecular Weight (g/mol) 343.20
MDL Number MFCD00237571
InChI Key ULNOXUAEIPUJMK-NSHDSACASA-M
Synonym s-n-boc-4-bromophenylalanine, boc-l-4-bromophenylalanine, boc-phe 4-br-oh, boc-4-bromo-l-phenylalanine, boc-l-4-bromophe, boc-p-bromo-phe-oh, boc-4'-bromo-l-phe, n-boc-4-bromo-l-phenylalanine, s-3-4-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-l-4-bromophenylalnine
PubChem CID 2734477
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Br)C=C1)C([O-])=O
Specifications
Melting Point 118°C to 120°C
Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-3-(4-bromophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Formula Weight 344.2
Percent Purity 98%
Chemical Name or Material N-Boc-4-bromo-L-phenylalanine