missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-bromo-L-phenylalanine, 95%

Product Code. 15498507
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15498507

Brand: Thermo Scientific Alfa Aesar H52024.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82278-73-7 | C14H18BrNO4 | 344.205 g/mol

N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82278-73-7
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD01317710
InChI Key FBUDYESOPLBQIR-NSHDSACASA-N
Synonym s-n-boc-3-bromophenylalanine, boc-l-3-bromophenylalanine, boc-3-bromo-l-phenylalanine, boc-phe 3-br-oh, boc-l-3-br-phe-oh, boc-l-3-bromophe, n-boc-3-bromo-l-phenylalanine, s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-3-bromophenyl propanoic acid
PubChem CID 2734476
IUPAC Name (2S)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 344.2
Percent Purity 95%
Chemical Name or Material N-Boc-3-bromo-L-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.