Learn More
N-Boc-1,3-propanediamine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-Boc-1,3-propanediamine |
| CAS | 75178-96-0 |
| Molecular Formula | C8H18N2O2 |
| Quantity | 5 g |
| Synonym | 1-Boc-amino-1,3-propanediamine, 1,1-Dimethylethyl (3-aminopropyl)carbamate, 1-[(tert-Butoxycarbonyl)amino]-3-aminopropane, N-BOC-1,3-diaminopropane, 1,1-Dimethylethyl N-[2-[2-(2-bromoethoxy)ethoxy]ethyl]carbamate, 2-[2-[2-(tert-Butoxycarbonylamino)ethoxy]ethoxy]ethyl Bromide, tert-Butyl (2-(2-(2-bromoethoxy)ethoxy)ethyl)carbamate |
| InChI Formula | InChI=1S/C8H18N2O2/c1-8(2,3)12-7(11)10-6-4-5-9/h4-6,9H2,1-3H3,(H,10,11) |
| SMILES | CC(C)(C)OC(=O)NCCCN |
| IUPAC Name | tert-butyl N-(3-aminopropyl)carbamate |
| Molecular Weight (g/mol) | 174.24 |
| Formula Weight | 174.1368 |