missing translation for 'onlineSavingsMsg'
Learn More

N-Biotinylcaproylaminoethyl Methanethiosulfonate, TRC

Product Code. 30346272
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346272 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346272 Supplier Toronto Research Chemicals Supplier No. TRCB39575010MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Biotinylcaproylaminoethyl Methanethiosulfonate
CAS 353754-95-7
Molecular Formula C19 H34 N4 O5 S3
Quantity 10 mg
InChI Formula InChI=1S/C19H34N4O5S3/c1-31(27,28)30-12-11-21-17(25)8-3-2-6-10-20-16(24)9-5-4-7-15-18-14(13-29-15)22-19(26)23-18/h14-15,18H,2-13H2,1H3,(H,20,24)(H,21,25)(H2,22,23,26)/t14-,15-,18-/m0/s1
SMILES CS(=O)(=O)SCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
IUPAC Name 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-N-(2-methylsulfonylsulfanylethyl)hexanamide
Molecular Weight (g/mol) 494.69
Formula Weight 494.1691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.