missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl L-Threonine Amide, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyloxycarbonyl Threonine Amide |
| CAS | 49705-98-8 |
| Molecular Formula | C12 H16 N2 O4 |
| Quantity | 500 mg |
| Synonym | Carbamic acid, N-[(1S,2R)-1-(aminocarbonyl)-2-hydroxypropyl]-, phenylmethyl ester, Carbamic acid, [(1S,2R)-1-(aminocarbonyl)-2-hydroxypropyl]-, phenylmethyl ester (9CI), Carbamic acid, [1-(aminocarbonyl)-2-hydroxypropyl]-, phenylmethyl ester, [R-(R*,S*)]-, N-(Benzyloxycarbonyl)-L-threonine amide, N-Benzyloxycarbonylthreonine amide |
| InChI Formula | InChI=1S/C12H16N2O4/c1-8(15)10(11(13)16)14-12(17)18-7-9-5-3-2-4-6-9/h2-6,8,10,15H,7H2,1H3,(H2,13,16)(H,14,17)/t8-,10+/m1/s1 |
| SMILES | C[C@@H](O)[C@H](NC(=O)OCc1ccccc1)C(=O)N |
| IUPAC Name | benzyl N-[(2S,3R)-1-amino-3-hydroxy-1-oxobutan-2-yl]carbamate |
| Molecular Weight (g/mol) | 252.27 |
| Formula Weight | 252.111 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?