Learn More
N-Benzyloxycarbonyl-L-glutamic acid, 99%
Product Code. 11483268
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11483268
Brand: Thermo Scientific Alfa Aesar A17686.18
Request Bulk or Custom Format
CAS: 1155-62-0 | C13H15NO6 | 281.264 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1155-62-0
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00002801
InChI Key PVFCXMDXBIEMQG-JTQLQIEISA-N
Synonym n-cbz-l-glutamic acid, z-glu-oh, n-benzyloxycarbonyl-l-glutamic acid, carbobenzoxy-l-glutamic acid, n-carbobenzoxy-l-glutamic acid, s-2-benzyloxy carbonyl amino pentanedioic acid, n-benzyloxycarbonylglutamic acid, n-carbobenzyloxy-l-glutamic acid, z-l-glu-oh
PubChem CID 70866
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCC(=O)O)C(=O)O
Specifications
Melting Point 108°C to 116°C
Quantity 50 g
Beilstein 2061272
Optical Rotation −7.5° (c=4 in Acetic Acid)
Formula Weight 281.27
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid