Learn More
Alfa Aesar™ N-Benzylidene-1,1-dimethyl-2-(4-fluorophenyl)ethylamine, 97%
Product Code. 15460847
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15460847 Supplier Alfa Aesar™ Supplier No. L17304.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 4117-09-3
Melting Point 43°C to 45°C
Boiling Point 135°C (0.3 mmHg)
Molecular Formula C7H13Br
MDL Number MFCD00151999
Quantity 5 g
Beilstein 2940979
Synonym 7-bromo-1-heptene, 1-heptene, 7-bromo, bromo-7 heptene-1, 7-bromo-heptene, 7-bromo-l-heptene, 1-bromo-6-heptene, acmc-2097en, ksc235k6r
InChI Key GNYDYUQVALBGGZ-UHFFFAOYSA-N
SMILES C=CCCCCCBr
IUPAC Name 7-bromohept-1-ene
Molecular Weight (g/mol) 177.09
PubChem CID 524450
Formula Weight 255.34
Percent Purity 97%
Chemical Name or Material N-Benzylidene-1,1-dimethyl-2-(4-fluorophenyl)ethylamine