Learn More
N-Benzyl Salbutamol Acetonide, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyl Salbutamol Acetonide |
| CAS | 1797114-57-8 |
| Molecular Formula | C23H31NO3 |
| Quantity | 25 mg |
| Synonym | α-[[(1,1-Dimethylethyl)(phenylmethyl)amino]methyl]-2,2-dimethyl-4H-1,3-benzodioxin-6-methanol, N-Benzyl Salbutamol Acetonide |
| InChI Formula | InChI=1S/C23H31NO3/c1-22(2,3)24(14-17-9-7-6-8-10-17)15-20(25)18-11-12-21-19(13-18)16-26-23(4,5)27-21/h6-13,20,25H,14-16H2,1-5H3 |
| SMILES | CC(C)(C)N(CC(O)c1ccc2OC(C)(C)OCc2c1)Cc3ccccc3 |
| IUPAC Name | 2-[benzyl(tert-butyl)amino]-1-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)ethanol |
| Molecular Weight (g/mol) | 369.4971 |
| Formula Weight | 369.2304 |