Learn More
N-Benzyl-(S)-proline, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Benzyl-(S)-proline |
| CAS | 31795-93-4 |
| Molecular Formula | C12 H15 N O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C12H15NO2/c14-12(15)11-7-4-8-13(11)9-10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9H2,(H,14,15)/t11-/m0/s1 |
| SMILES | OC(=O)[C@@H]1CCCN1Cc2ccccc2 |
| IUPAC Name | (2S)-1-benzylpyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 205.25 |
| Formula Weight | 205.1103 |