Learn More
N-Benzyl Carvedilol-d5, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | N-Benzyl Carvedilol-d5 |
| CAS | 1329792-68-8 |
| Molecular Formula | C31H27D5N2O4 |
| Quantity | 25 mg |
| Synonym | 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl][(phenyl-d5)methyl)]amino]-2-propanol |
| InChI Formula | InChI=1S/C31H32N2O4/c1-35-28-15-7-8-16-29(28)36-19-18-33(20-23-10-3-2-4-11-23)21-24(34)22-37-30-17-9-14-27-31(30)25-12-5-6-13-26(25)32-27/h2-17,24,32,34H,18-22H2,1H3/i2D,3D,4D,10D,11D |
| SMILES | [2H]c1c([2H])c([2H])c(CN(CCOc2ccccc2OC)CC(O)COc3cccc4[nH]c5ccccc5c34)c([2H])c1[2H] |
| IUPAC Name | 1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]amino]propan-2-ol |
| Molecular Weight (g/mol) | 501.63 |
| Formula Weight | 501.2676 |