Learn More
N-Benzyl-bis(2-chloroethyl)amine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-Benzyl-bis(2-chloroethyl)amine Hydrochloride |
| CAS | 10429-82-0 |
| Molecular Formula | C11H15NCl2 . HCl |
| Quantity | 500 mg |
| Synonym | Benzylbis(2-chloroethyl)amine hydrochloride |
| InChI Formula | InChI=1S/C11H15Cl2N.ClH/c12-6-8-14(9-7-13)10-11-4-2-1-3-5-11;/h1-5H,6-10H2;1H |
| SMILES | Cl.ClCCN(CCCl)Cc1ccccc1 |
| IUPAC Name | N-benzyl-2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 268.61 |
| Formula Weight | 267.0348 |