Learn More
N-Benzyl-3,3'-iminodipropionic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | N-Benzyl-3,3'-iminodipropionic Acid |
| CAS | 6405-28-3 |
| Molecular Formula | C13H17NO4 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C13H17NO4/c15-12(16)6-8-14(9-7-13(17)18)10-11-4-2-1-3-5-11/h1-5H,6-10H2,(H,15,16)(H,17,18) |
| SMILES | OC(=O)CCN(CCC(=O)O)Cc1ccccc1 |
| IUPAC Name | 3-[benzyl(2-carboxyethyl)amino]propanoic acid |
| Molecular Weight (g/mol) | 251.28 |
| Formula Weight | 251.1158 |