missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-DL-alanine, 97+%

Product Code. p-7046088
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337886 10 g 10g
11347886 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337886 Supplier Thermo Scientific Alfa Aesar Supplier No. L13100.09
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1205-02-3 | C10H11NO3 | 193.20 g/mol

N-Benzoyl-DL-alanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-DL-alanine is used as pharmaceutical intermediate.

Solubility
Very slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1205-02-3
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00020393
InChI Key UAQVHNZEONHPQG-UHFFFAOYNA-N
Synonym n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine
PubChem CID 71002
ChEBI CHEBI:71167
IUPAC Name 2-benzamidopropanoic acid
SMILES CC(NC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 163°C to 167°C
Beilstein 3201778
Solubility Information Very slightly soluble in water.
Quantity 10 g
Formula Weight 193.2
Percent Purity ≥97%
Chemical Name or Material N-Benzoyl-DL-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.