Learn More
N-Acetyl-S-(2-carbamoylethyl)-L-cysteine, CDN
Product Code. 30321238
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Product Code. 30321238
Brand: CDN CDNX61880.05G
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl-S-(carbamoylethyl)-L-cysteine
CAS 81690-92-8
Molecular Formula C8 H14 N2 O4 S
InChI Formula InChI=1 S/C8H14N2O4S/c1-5(11)10-6(8(13)14)4-15-3-2-7(9)12/h6H,2-4H2,1H3,(H2,9,12)(H,10,11)(H,13,14)/t6-/m0/s1
SMILES CC(=O)N[C@@H](CSCCC(=O)N)C(=O)O
IUPAC Name (2 R)-2-acetamido-3-(3-amino-3-oxopropyl)sulfanylpropanoic acid
Molecular Weight (g/mol) 234.27
Quantity 0.05 g
Formula Weight 234.0674 g/mol
Percent Purity min 98% chemical purity
Shipping Condition Room Temperature