missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-acetyl-L-cysteine, >96%, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Specifications
Specifications
| Color | White |
| Assay Percent Range | >96% |
| Density | 1.295g/cm3 (L) |
| Quantity | 1 kg |
| Formula Weight | 163.2 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-acetyl-L-cysteine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction