missing translation for 'onlineSavingsMsg'
Learn More

Melford n-Acetyl-L-cysteine

Product Code. 30017930
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
This item is not returnable. View return policy

Product Code. 30017930

Brand: Melford A10040100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Increases cellular pools of free radical scavenger.

Increases cellular pools of free radical scavenger. Prevents apoptotic death of neuronal cells. Induces apoptosis in human and rat smooth muscle cells. Pyrogen free.

Specifications

Chemical Name or Material n-Acetyl-L-cysteine
CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.2
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.