Learn More
N-Acetyl-DL-Valine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-DL-valine |
| CAS | 3067-19-4 |
| Molecular Formula | C7 H13 N O3 |
| Quantity | 500 mg |
| Synonym | Valine, N-acetyl-, DL-Valine, N-acetyl-, Valine, N-acetyl-, DL- (8CI), N-Acetylvaline, 2-Acetamido-3-methylbutanoic acid, Acetyl-DL-valine, DL-N-Acetylvaline, N-Acetyl-DL-valine, NSC 122016 |
| InChI Formula | InChI=1S/C7H13NO3/c1-4(2)6(7(10)11)8-5(3)9/h4,6H,1-3H3,(H,8,9)(H,10,11) |
| SMILES | CC(C)C(NC(=O)C)C(=O)O |
| IUPAC Name | 2-acetamido-3-methylbutanoic acid |
| Molecular Weight (g/mol) | 159.183 |
| Formula Weight | 159.0895 |