Learn More
N-Acetyl-3-iodo-L-tyrosine Amide, TRC
Product Code. 30320379
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320379 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30320379 Supplier Toronto Research Chemicals Supplier No. TRCA179410500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl-3-iodo-L-tyrosine, Amide
CAS 59302-19-1
Molecular Formula C11 H13 I N2 O3
Quantity 500 mg
InChI Formula InChI=1S/C11H13IN2O3/c1-6(15)14-9(11(13)17)5-7-2-3-10(16)8(12)4-7/h2-4,9,16H,5H2,1H3,(H2,13,17)(H,14,15)/t9-/m0/s1
SMILES CC(=O)N[C@@H](Cc1ccc(O)c(I)c1)C(=O)N
IUPAC Name (2S)-2-acetamido-3-(4-hydroxy-3-iodophenyl)propanamide
Molecular Weight (g/mol) 348.14
Formula Weight 347.9971