Learn More
N-Acetonylpyridinium chloride, 97%
Product Code. 11458636
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458636 5 g 5g
11468636 25 g 25g
11478636 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11458636 Supplier Thermo Scientific Alfa Aesar Supplier No. A16015.06
Request Bulk or Custom Format
CAS: 42508-60-1 | C8H10ClNO | 171.62 g/mol
N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from water and humidity.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42508-60-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00031949
InChI Key JHYYWKUENNZTMD-UHFFFAOYSA-M
Synonym 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride
PubChem CID 2734121
SMILES [Cl-].CC(=O)C[N+]1=CC=CC=C1
Specifications
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name 1-(2-oxopropyl)pyridin-1-ium chloride
Formula Weight 171.63
Percent Purity 97%
Chemical Name or Material N-Acetonylpyridinium chloride