Learn More
Macherey-Nagel™ Septa for Type N 8 Screw Caps
Product Code. p-8012171
Click to view available options
Color:
Blue, White
Red, White
White
Material:
PTFE Virginal
PTFE, Silicone
Pre-slit:
No
Yes
Unit Size:
Pack of 100
Product Code. 10095671
Brand: Macherey-Nagel™ 70261
Only null left
Add to basket
Add to basket
Maintain the integrity of the sample, while allowing syringe penetration. Macherey-Nagel™ Septa for Type N 8 Screw Caps provide an air-tight seal that prevents contamination. Available in a variety of materials. Polypropylene screw caps with a center hole sold separately.
  • PTFE septa is chemically resistant and allows puncturing
  • Silicone lining allows repeated puncturing by preventing coring damage
  • Slitted septa require less pressure to penetrate, preventing needle damage
  • Packaged as 100 pieces (septa only) in a polyethylene bag
  • Linerless black polypropylene screw caps with a center hole for type N8 vials are sold separately (mfr. no. 70249)
Specifications
Description Hardness: 53° Shore D
Diameter (Metric) Cap 8 mm
Color White
Material PTFE Virginal
Pre-slit No
Septum Septa N 8
For Use With (Application) Chromatography (GC, HPLC)
Style Round
Quantity 100 Pk.
Thickness (Metric) 0.25 mm
Closure Size N8