Learn More
Macherey-Nagel™ N 8 Aluminum Crimp Caps
Click to view available options
Thickness (Metric):
0.25 mm
1 mm
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Silver |
| Material | Aluminum, PTFE |
| Septum | PTFE Virginal/White |
| Quantity | 100 Pk. |
| Thickness (Metric) | 0.25 mm |