missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[4-[(Trifluoroacetyl)amino]benzoyl]-L-glutamic Acid Dimethyl Ester-13C5, TRC
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | N-[4-[(Trifluoroacetyl)amino]benzoyl]-L-glutamic Acid Dimethyl Ester-13C5 |
| Molecular Formula | 13C5 C11 H17 F3 N2 O6 |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C16H17F3N2O6/c1-26-12(22)8-7-11(14(24)27-2)21-13(23)9-3-5-10(6-4-9)20-15(25)16(17,18)19/h3-6,11H,7-8H2,1-2H3,(H,20,25)(H,21,23)/t11-/m0/s1/i7+1,8+1,11+1,12+1,14+1 |
| SMILES | CO[13C](=O)[13CH2][13CH2][13C@H](NC(=O)c1ccc(NC(=O)C(F)(F)F)cc1)[13C](=O)OC |
| IUPAC Name | dimethyl (2S)-2-[[4-[(2,2,2-trifluoroacetyl)amino]benzoyl]amino](1,2,3,4,5-13C5)pentanedioate |
| Molecular Weight (g/mol) | 395.274 |
| Formula Weight | 395.121 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction