Learn More
N-[4-(2-Bromoethyl)phenyl]methanesulfonamide, TRC
Product Code. 30335970
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335970 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30335970 Supplier Toronto Research Chemicals Supplier No. TRCB68812450MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material N-[4-(2-Bromoethyl)phenyl]methanesulfonamide
CAS 90485-25-9
Molecular Formula C9H12BrNO2S
Quantity 50 mg
InChI Formula InChI=1S/C9H12BrNO2S/c1-14(12,13)11-9-4-2-8(3-5-9)6-7-10/h2-5,11H,6-7H2,1H3
SMILES CS(=O)(=O)Nc1ccc(CCBr)cc1
IUPAC Name N-[4-(2-bromoethyl)phenyl]methanesulfonamide
Molecular Weight (g/mol) 278.17
Formula Weight 276.9772