Learn More
N-[(2R,4R,5S)-5-[[(1,1-Dimethylethoxy)carbonyl]amino]-4-hydroxy-1-oxo-6-phenyl-2-(phenylmethyl)hexyl]-L-leucyl-L-phenylalaninamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-((2R,4R,5S)-5-(((1,1-Dimethylethoxy)carbonyl)amino)-4-hydroxy-1-oxo-6-phenyl-2-(phenylmethyl)hexyl)-L-leucyl-L-phenylalaninamide |
| CAS | 292632-98-5 |
| Molecular Formula | C39H52N4O6 |
| Quantity | 5 mg |
| Synonym | 36: PN: WO2005049027 PAGE: 99 claimed sequence, L 458, L 685458, gamma-Secretase Inhibitor X |
| InChI Formula | InChI=1S/C39H52N4O6/c1-26(2)21-33(37(47)41-32(35(40)45)24-29-19-13-8-14-20-29)42-36(46)30(22-27-15-9-6-10-16-27)25-34(44)31(23-28-17-11-7-12-18-28)43-38(48)49-39(3,4)5/h6-20,26,30-34,44H,21-25H2,1-5H3,(H2,40,45)(H,41,47)(H,42,46)(H,43,48)/t30-,31+,32+,33+,34-/m1/s1 |
| SMILES | CC(C)C[C@H](NC(=O)[C@@H](C[C@@H](O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C)Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N |
| IUPAC Name | tert-butyl N-[(2S,3R,5R)-6-[[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-5-benzyl-3-hydroxy-6-oxo-1-phenylhexan-2-yl]carbamate |
| Molecular Weight (g/mol) | 672.85 |
| Formula Weight | 672.3887 |