Learn More
N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid tert-Butyl Ester, TRC
Product Code. 30393371
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393371 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30393371 Supplier Toronto Research Chemicals Supplier No. TRCH942845100MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid tert-Butyl Ester
CAS 1009093-14-4
Molecular Formula C14H21NO4
Quantity 100 mg
Synonym N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C14H21NO4/c1-14(2,3)19-13(18)15-11(9-16)12(17)10-7-5-4-6-8-10/h4-8,11-12,16-17H,9H2,1-3H3,(H,15,18)/t11-,12-/m1/s1
SMILES CC(C)(C)OC(=O)N[C@H](CO)[C@H](O)c1ccccc1
IUPAC Name tert-butyl N-[(1R,2R)-1,3-dihydroxy-1-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol) 267.32
Formula Weight 267.1471