missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]hydroxyphosphinyl]-D-alanine 1-Methylethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-((((1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy)methyl)hydroxyphosphinyl)-D-alanine 1-Methylethyl Ester |
| CAS | 376633-26-0 |
| Molecular Formula | C15 H25 N6 O5 P |
| Quantity | 5 mg |
| Synonym | GS 7485 |
| InChI Formula | InChI=1S/C15H25N6O5P/c1-9(2)26-15(22)11(4)20-27(23,24)8-25-10(3)5-21-7-19-12-13(16)17-6-18-14(12)21/h6-7,9-11H,5,8H2,1-4H3,(H2,16,17,18)(H2,20,23,24)/t10-,11-/m1/s1 |
| SMILES | CC(C)OC(=O)[C@@H](C)NP(=O)(O)CO[C@H](C)Cn1cnc2c(N)ncnc12 |
| IUPAC Name | [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-N-[(2R)-1-oxo-1-propan-2-yloxypropan-2-yl]phosphonamidic acid |
| Molecular Weight (g/mol) | 400.37 |
| Formula Weight | 400.162 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?