missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Deoxy-D-fructos-1-yl)-L-methionine, TRC

Product Code. 30364329
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364329 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364329 Supplier Toronto Research Chemicals Supplier No. TRCD23609010MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(1-Deoxy-D-fructos-1-yl)-L-methionine
CAS 20638-92-0
Molecular Formula C11 H21 N O7 S
Quantity 10 mg
Synonym (S)-4-(Methylthio)-2-(((3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexyl)amino)butanoic Acid
InChI Formula InChI=1S/C11H21NO7S/c1-20-3-2-6(11(18)19)12-4-7(14)9(16)10(17)8(15)5-13/h6,8-10,12-13,15-17H,2-5H2,1H3,(H,18,19)/t6-,8+,9+,10+/m0/s1
SMILES CSCC[C@H](NCC(=O)[C@@H](O)[C@H](O)[C@H](O)CO)C(=O)O
IUPAC Name (2S)-4-methylsulfanyl-2-[[(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexyl]amino]butanoic acid
Molecular Weight (g/mol) 311.352
Formula Weight 311.104
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.