missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MyoCult™ Differentiation Kit (Human)
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
Description
- MyoCult™ Differentiation Kit (Human) includes MyoCult™ Differentiation Medium and Animal Component-Free (ACF) Cell Attachment Substrate
- MyoCult™ Differentiation Medium is a serum-based medium specifically formulated for the in vitro differentiation of human skeletal muscle progenitor cells (myoblasts) into myotubes
- MyoCult™ Differentiation Medium is designed for use with the ACF Cell Attachment Substrate, which has been specifically tested for its ability to support optimal adherence and myotube morphology
- This medium is suitable for the differentiation of myogenic progenitor cells previously expanded in serum-containing or serum-free media (e.g. using the MyoCult™-SF Expansion Kit #05980)
Specifications
Specifications
| Cell Type | Myogenic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 200 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction