missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MYL6 (Human) Recombinant Protein
Human MYL6 full-length ORF ( NP_066299.2, 1 a.a. - 151 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004637-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MCDFTEDQTAEFKEAFQLFDRTGDGKILYSQCGDVMRALGQNPTNAEVLKVLGNPKSDEMNVKVLDFEHFLPMLQTVAKNKDQGTYEDYVEGLRVFDKEGNGTVMGAEIRHVLVTLGEKMTEEEVEMLVAGHEDSNGCINYEAFVRHILSG
Specifications
NP_066299.2 | |
myosin, light chain 6, alkali, smooth muscle and non-muscle | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MYL6 | |
GST |
4637 | |
Wheat germ expression system | |
10 μg | |
ESMLC, LC17-GI, LC17-NM, LC17A, LC17B, MLC1SM, MLC3NM, MLC3SM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |