Description
- A fast and cost-effective way to verify the absence of mycoplasma in your cell lines
- This service leverages the MycoSEQ Mycoplasma Detection Kit (4460623), a real-time PCR assay designed and validated to meet the requirements of European Pharmacopeia Chapter 2.6.7
- The primers were designed using a proprietary bioinformatics pipeline to detect >90 mycoplasma species with high specificity and no cross-reactivity to closely related bacterial species
The MycoSeq Assay Service offers:
- Fast data delivery – turnaround time of four weeks or less
- Convenient sample prep – just send a frozen cell pellet
- High sensitivity – demonstrated by internal and external validations to detect 10 cfu/mL
- High specificity – includes proprietary Discriminatory Positive Control to differentiate true positive from any potential cross-contamination positives
- Exceptional support – a dedicated project manager for frequent project communication
Specifications
Specifications
| Quantity | 1 sample |
| Duration | 3 Weeks |
| Product Type | Assay Service |
| Product Line | Applied Biosystems, QuantStudio |
| Field | Research Use Only |
| Target Audience | Cell Line Developers |
| Species | All |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?