Learn More
MP Biomedicals™ Mycoplasma Hoechst Stain Kit
Product Code. p-7236586 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
Product Code. 11495065
Brand: MP Biomedicals™ 093030000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The Mycoplasma Hoechst Stain Kit is designed for rapid in situ detection of mycoplasma, and other prokaryotic organisms, in cell cultures in less than two hours.
  • Uses DNA fluorochrome staining technology with Hoechst Stain 33258
  • For the in situ detection of mycoplasma, bacteria, yeast, and fungi in cell cultures
  • Testing is rapid (less than 2 hours)and economical (100 tests per kit)
  • Non-nuclear staining is readily apparent, with contaminants standing out boldly against a black background
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 15° to 30°C
Product Type Mycoplasma Detection
Quantity 100 Tests
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.