missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mumps Virus Native Protein

Product Code. 18688087 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 18688087

Brand: Novus Biologicals™ NBP262509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Formulation Inactivated Mumps virus - liquid. Culture medium 1969 B
Quantity 1 mL
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Inhibitor Properties Antigenic activity is 51.1% of internal reference batch
Protein Mumps Virus
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.