Learn More
BOLA Multiple Distributors for Bottles
Product Code. p-200110002
Click to view available options
Thread Size (Metric):
GL 18
GL 25
GL 32
GL 40
GL 49
Unit Size:
Each
Product Code. 10769303
Brand: BOLA D61908
Only null left
Add to basket
Add to basket
Multiple Distributors for Bottles PTFE. Screw cap
Black screw cap made of PPS for thread according to chart. Body made of PTFE.
Specifications
Color White
Diameter (Metric) Outer Neck 3 x 6 mm
Sterility Non-sterile
Thread Size (Metric) GL 25
Neck Finish 61 mm
Certifications/Compliance FDA
Disposable Reusable
Autoclavable Autoclavable