Learn More
Hanna Instruments™ Multiparameter Photometer
Specifications
Specifications
| Type | Multiparameter Photometer |
| Display Type | LCD |
| Connection Type | USB |
| For Use With (Application) | Wastewater Analysis |
| Voltage | 115/230 VAC |
| Height (Metric) | 110 mm |
| Length (Metric) | 235 mm |
| Light Source | Tungsten lamps with different narrow band interference filters |
| Weight (Metric) | 0.9 kg |
| Width (Metric) | 200 mm |