Learn More
Hanna Instruments™ Multiparameter Meter Hi 9829
Click to view available options
Includes:
4 m GPS Cable
Unit Size:
Each
Description
pH/ORP, with 4m GPS cable, Hanna Instruments
Specifications
Specifications
| Includes | 4 m GPS Cable |
| Product Type | ph/ORP Meter |