Learn More
Gestigkeit Harry Series heating bath, 4 units, 1L, 2000W, 230V
Brand:  Gestigkeit Harry SM4
Series heating bath, 4 units, 1L, 2000W, 230V
Product Code. 10016111
  • ONLINE EXCLUSIVE PRICE
    1753.50€
    1622.52€ / Each
    Save 130.98€
    7% Off
Only null left
Estimated Shipment: 20-02-2026