Learn More
Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories
Product Code. p-4528677 Shop All Thermo Scientific Products
Click to view available options
Description:
SMART 2 Long Standard, New RFID Tag
SMART 2 Standard, New RFID Tag
SMART+ Long Standard
SMART+ Small Metal Tip
SMART+ Small Plastic Tip
SMART+ Standard
Volume (Metric):
0.5 to 50 μL
5 to 2500 μL
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 15387400
Brand: Thermo Scientific™ N15137
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.
  • Designed for use with Multidrop Combi SMART+ dispensers
  • Equipped with radio frequency identification (RFID) tags to store lifetime dispensing data and display remaining usage time
  • Available in standard and small tube versions
  • Distinguished from normal cassettes by antenna tags
  • Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
  • Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tube Dispensing Cassette
Autoclavable Autoclavable (50 times at 121°C, 1 bar, 20 min.)
Description SMART 2 Standard, New RFID Tag
Material Polypropylene Tip, Silicone Tubing
Volume (Metric) 5 to 2500 μL
For Use With (Equipment) Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
Quantity 1 Ea.
Volume (Metric) Dead <7 mL
Diameter (Metric) Tubing: 1.3 mm; Tip: 0.5 mm