missing translation for 'onlineSavingsMsg'
Learn More

Mutexil™ Multi-Use Roll, Handtowel

Product Code. 15896761
Click to view available options
Packaging Quantity:
7 Pk. x 2/Bag, 50 Pk/Cs.
Unit Size:
Pack of 14
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.

Product Code. 15896761

Brand: Mutexil™ 9015214

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.

Convenient, absorbent, multi-use handtowel/wiper.

  • Multiple cleaning uses
  • Roll format
  • Absorbent paper construction

Specifications

Material Paper
Format Roll
For Use With (Application) Multi-use
Packaging Quantity 7 Pk. x 2/Bag, 50 Pk/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.