Learn More
Melford MUG
Product Code. 30018522
Click to view available options
Quantity:
100 mg
250 mg
1 g
25 g
Unit Size:
100mg
1g
250mg
25g
Product Code. 30018522
Brand: Melford M659000.1
Fluorescent substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene isolated from E.
Fluorescent substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene isolated from E. coli. Fluorescence assay allows quantitation of GUS activity in protein extracts at peak excitation of 365 nm (UV) and a peak emission of 455 nm (blue).
Specifications
Quantity 100 mg