Learn More
Thermo Scientific™ Mueller Hinton Agar
Product Code. p-4523375
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Unit Size:
Pack of 10
Product Code. 13235349
Brand: Thermo Scientific™ PO5007A
Only null left
Add to basket
Add to basket
Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.
The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.
Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance According to EUCAST and CLSI
Color Ivory, transparent
Content And Storage 2° to 12°C, in the dark
Description Mueller Hinton Agar
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 24 g ± 5 %
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.