Learn More
Brand™ Mucocit™ Disinfecting Detergents
Shop All Brand Products
Click to view available options
Volume (Metric):
2 L
Unit Size:
10 L
Specifications
Specifications
| Product Type | Mucocit-T Disinfecting Detergent |
| Form | Liquid |
| Appearance | Clear Blue |
| Composition | 100g Mucocit™ T Contain: 7g Didecyldimethylammoniumchloride, 4.5g Alkylpropylenediamino-1,5-Bi |
| Container Type | Bottle |
| For Use With (Application) | For Sensitive Instruments |
| Quantity | 1 Ea. |
| Volume (Metric) | 2 L |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : poly(oxy-1,2-ethandiyl)alpha-tridecyl-omega-hydroxyl branched, didecyldimethyl-ammonium chloride, te