Learn More
IKA™ MTT 100.10 Reusable Grinding Chamber
Product Code. 16298561 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
16298561 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16298561 Supplier IKA™ Supplier No. 0020019576
  • ONLINE EXCLUSIVE PRICE
    213.00€
    197.00€ / Pack of 10
    Save 16.00€
    7% Off
In Stock
Reusable grinding chamber equipped with a titanium beater.
  • With titanium beater
  • Used for heavy metal analysis
  • Chamber is made of transparent plastic
  • Allows grinding tests to be observed at any time
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Material Polypropylene
Packaging Quantity 10 Pk.
Weight (Metric) 0.516 kg