missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MTPN Recombinant Protein
Brand: Abnova™ H00136319-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH28093 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
38.72 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MCDKEFMWALKNGGLDEVKDYVAKGEDVNRTLEGGRKPLHYAADCGQLEILEFLLLKGADINAPDKHHITPLLSAVYEGHVSCVKLLLSKGADKTVKGPDGLTAFEATDNQAIKALLQ | |
FLJ31098/FLJ99857/GCDP/V-1 | |
MTPN | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
136319 | |
MTPN (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MTPN | |
Human | |
Recombinant | |
Solution |