Learn More
Carbolite™ MTF 12/25/400 Wire Wound Single Zone Tube Furnace
Product Code. 12039159
Click to view available options
Analysis Time:
Heat-up time: 25 min.
Unit Size:
Each
Product Code. 12039159
Brand: Carbolite™ M1225400230SN+&02301
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Uses wire element that is wound directly onto fixed integral ceramic work tube.
  • Simple and economical design
  • Integral wire wound work tube
  • Delayed start/process timer function as standard
  • Horizontal configuration mounted on control module
  • Carbolite Gero 301 controller, with single ramp to set-point and process timer
Specifications
Analysis Time Heat-up time: 25 min.
Controller Type Carbolite Gero 301
Temperature (Metric) 1100°C to 1200°C
Type Thermocouple Type: N
Power Consumption 1000 w
Dimensions (L x W x H) Exterior W x H: 450 x 375 mm
Depth (Metric) 375 mm
Weight (Metric) 15 kg
Wattage 200 w holding