Learn More
STEMCELL Technologies mTeSR™3D
Product Code. 15725593
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 15725593
Brand: STEMCELL Technologies 03950
Defined, serum-free media for suspension culture of human ES and iPS cells
  • mTeSR™3D was developed for expansion and scale-up of undifferentiated human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture
  • The novel fed-batch workflow saves time and media
  • Daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days
  • mTeSR™3D is a specialised and serum-free formulation
  • This culture system is compatible with a range of suspension culture vessels
Specifications
Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture, Expansion
Quantity 1 Kit
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.