missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MT1K (Human) Recombinant Protein
Human MT1K full-length ORF ( AAH28280, 1 a.a. - 61 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004499-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDPNCSCTTGVSCACTGSCKCKECKCTSCKKSCCSCCPVGCAKCAHGCVCKGTLENCSCCA
Specifications
AAH28280 | |
metallothionein 1M | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MT1M | |
GST |
4499 | |
Wheat germ expression system | |
25 μg | |
MGC118949, MGC40498, MT1, MT1K | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |