missing translation for 'onlineSavingsMsg'
Learn More

Corning™ MSCulture Max™- Supplement

Product Code. 31937649
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31937649 1 Ea. 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31937649 Supplier Corning™ Supplier No. 42000CR
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-performance expansion media for batch culture systems.

    The media increase final hMSC yields, streamlines culture, reduces in-process risk, and eliminates media exchanges, enabling higher cell expansion with less media and fewer media changes. The MSCulture Max media is available in 3 versions. Each media version is made up of a basal medium and a corresponding supplement
  • Standard Research-grade: Corning MSCulture Max Basal Medium [Cat. No. 42-010-CV] and Corning MSCulture Max Supplement [Cat. No. 42-000-CR]
  • Xeno-free Research-grade: Corning MSCulture Max Basal Medium [Cat. No. 42-010-CV] and MSCulture Max-XF Supplement [Cat. No. 42-100-CR]
  • Xeno-free GMP-grade for Bioproduction: Corning MSCulture Max Basal Medium [Cat. No. 42-010-CV] and Corning MSCulture Max-GMP Supplement [Cat. No. 42-200-CR]
  • Features:
    • No media exchanges required
    • No surface coating required
    • Unparalleled productivity: Achieves over 10-fold hMSC expansion within 4-5 days without media changes, significantly reducing time and labor costs
    • Transition-friendly formulation: Supports translation from research into clinical manufacturing through research and xeno-free formats. Xeno-free MSCulture Max media is also available in a GMP-grade version.
    • Scalable and efficient bioprocessing: Enables high-volume cell production with minimal media consumption
TRUSTED_SUSTAINABILITY

Specifications

Classification Serum-free
Form Liquid
Product Type Cell Culture Media
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives None
Quantity 1 Ea.
Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic), Adherent Cell Culture, Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.